L(-)-Tryptophan ≥99%, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
L-α-Amino-3-indolepropionic acid
, (S)-2-Amino-3-(3-indolyl)propionic acid
, H-Trp-OH
, L-Tryptophan
, Tryptophan
L-Tryptophan is an amino acid precursor of serotonin and melatonin.
Chloride ≤0.05%
sulphate ≤0.03%
Iron ≤0.003%
It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.
It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.
Soluble in 1N HCl (10 mg/mL - clear, colourless solution).
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 |
Specification Test Results
Synonyms | L-2-amino-3-indolepropionic acid; Trp; W; (S)-2-Amino-3-(3-indolyl)propionic acid |
Form | White powder |
Storage | Store at Room temperature(15-30 °C). |
Heavy Metals | ≤0.0015% |
Moisture Content | ≤0.3% |
Optical Rotation | -30° ± 10° (c=1, water) |
pH | 5.5 - 7.0 (1% aq soln) |
Residue on Ignition | ≤0.1% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...