Taurine ≥99%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-Aminoethanesulphonic acid, 2-Aminoethanesulfonic acid

0210300310 0210300380 0210300390 0210300391
IC10300310EA 57.3 USD
IC10300310 IC10300380 IC10300390 IC10300391
Taurine ≥99%, white crystalline powder
Taurine

Taurine is a sulphonated amino acid that occurs abundantly in vivo, and is found in oxen lung, shark blood, mussels and oysters. It is produced through the metabolism of cysteine via the formation of the intermediates cysteinesulphinate and hypotaurine.


It has been proposed to play important roles in biological functions such as regulating intracellular calcium, osmoregulation, brain development (particularly for cerebellum and retina cells), and in the enhancement of bile flow and cholesterol clearance by the liver. Taurine may be conjugated to bile acids and secreted in the duodenum.


It is an inhibitor of lipid peroxidation. Inhibits diethyl nitrosamine and phenobarbital-induced hepatocarcinogenesis. It has been used as a sulphur source in examining the gene expression profile of transporters in Bacillus subtilis via DNA array analysis. A report on the use of taurine as a sole sulphur source by several strains of E. coli has been published. The cytoprotective role of taurine has been investigated in cell culture, and its uptake in lactating porcine mammary tissue has been studied.


Non-selective endogenous agonist at glycine receptors. Conditionally essential sulphonated amino acid which modulates apoptosis in some cells; functions in many metabolic activities; a product of methionine and cysteine metabolism.


Soluble in water (50 mg/mL-clear, colorless solution).


Taurine is a sulfonated amino acid that occurs abundantly in vivo, and is found in oxen lung, shark blood, mussels and oysters. It is produced through the metabolism of cysteine via the formation of the intermediates cysteinesulfinate and hypotaurine. Taurine has been proposed to play important roles in biological functions such as regulating intracellular calcium, osmoregulation, brain development (particularly for cerebellum and retina cells), and in the enhancement of bile flow and cholesterol clearance by the liver. Taurine may be conjugated to bile acids and secreted in the duodenum.

Formula: C₂H₇NO₃S
MDL Number: MFCD00008197
CAS Number: 107-35-7

Order Now

Specification Test Results

Synonyms 2-Aminoethanesulfonic acid
Form White Crystalline Powder
Storage Store at Room Temperature(15-30 °C).
Loss on Drying ≤0.5%
pka pKa (-SO3H): 1.5; pKa (-NH3): 9.06, 8.74(Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR