Tin(II) chloride dihydrate, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Stannous chloride dihydrate

0210294980 0210294990 0210294991
IC10294980EA 106.9 USD
IC10294980 IC10294990 IC10294991
Tin(II) chloride dihydrate, white crystalline powder
Tin (II) chloride dihydrate

Soluble in hydrochloride.

Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623
Melting Pt: 246
MDL Number: MFCD00149863
CAS Number: 10025-69-1
UN: 3260
ADR: 8 III
Merck Index: 13,08861
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5151cc27e385537403bd1f46c43c835430850da5&vid=5151cc27e385537403bd1f46c43c835430850da5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR