Sodium chloride 99,0-100,5, powder USP

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Saline, NaCl, Sea salt, Halite

02102892.5 0210289201 0210289205 0210289210
IC102892.5EA 87.95 USD
IC102892.5 IC10289201 IC10289205 IC10289210
Sodium chloride 99,0-100,5, powder USP
Sodium chloride

Soluble in water (357 mg/mL at 25 °C; 384 mg/mL at 100 °C), glycerol (100 mg/mL); very slightly soluble in ethanol. Solubility in water is decreased by HCl; almost insoluble in concentrated HCl. Aqueous solutions are at a neutral pH (6.7 - 7.3). Aqueous solutions of sodium chloride are stable at room temperature.


Aluminum= ≤0.2 µg/g; Magnesium and Alkaline-earth Metals=≤0.01% (as Ca); Arsenic=≤1 µg/g; Iron=≤2 µg/g; Limit of Phosphates =≤0.0025%; Limit of Potassium =≤0.05%.


Sodium chloride is used in a wide variety of biochemical applications, including intravenous fluids (normal saline is 0.9% w/v in water), density gradients, a diluent to increase ionic strength in buffers or culture media and in salt-out procedures in the isolation of DNA. It has also been used in high concentrations for preservation of foods since bacteria cannot grow in high salt conditions. A salt-ice mixture in the ratio of approximately 33 g sodium chloride to 100 g ice (at -1°C) will drop in temperature to as low as -21°C, depending on the rate of stirring and the size of the ice chunks.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e63174fdceac5a60d255d7d63517ed46cad4ab3&vid=4e63174fdceac5a60d255d7d63517ed46cad4ab3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms Salt; Dendritis; Halite; Purex; Rock salt; saline; sterling; table salt; top flake
Form Powder
Storage Store at Room Temperature (15-30 °C)
Heavy Metals ≤5 ppm
Loss on Drying ≤0.5%
Sterilization of Solutions It can be sterilized by autoclaving or sterile filtering.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR