L(+)-Serine ≥99%, free flowing white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
L-serine is used in the study of influence of pulsed applications of serine on polyglucose synthesis by Fusobacterium nucleatum cultures. Supplementation of minimal media with serine has been shown to restore growth in Escherichia coli that lack the PII and GlnK genes.
Precursor of glycine by serine hydroxymethyltransferase.
Soluble in water (50 mg/mL); insoluble in alcohol and ether.
L-serine is used in the study of influence of pulsed applications of serine on polyglucose synthesis by Fusobacterium nucleatum cultures. Supplementation of minimal media with serine has been shown to restore growth in Escherichia coli that lack the PII and GlnK genes.
Formula:
C₃H₇NO₃ Melting Pt: 222 °C (dec., lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00064224 CAS Number: 56-45-1 |
Specification Test Results
Synonyms | L-2-Amino-3-hydroxypropionic acid; Ser; S; (S)-2-amino-3-hydroxypropionic acid; β-hydroxyalanine; α-amino-β-hydroxypropionic acid |
Form | Free Flowing White Powder |
Storage | Store at Room temperature(15-30 °C). |
Heavy Metals | ≤20 ppm(as Pb) |
Loss on Drying | ≤0.5% |
Optical Rotation | +10° to +20° (c = 10, 2N HCl) |
pka | 2.19 (-COOH), 9.21 (-NH2)(Lit.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...