Phenolphthalein ≥98%, white powder USP, NF

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

0210260880 0210260883 0210260890
IC10260880EA 59.99 USD
IC10260880 IC10260883 IC10260890
Phenolphthalein ≥98%, white powder USP, NF
Phenolphthalein

Soluble in alcohol, ether, methanol (10 mg/mL - clear, yellow solution), ethanol(50 mg/mL), acetone; very slightly soluble in chloroform; almost insoluble in water.


Soluble in alcohol, ether, methanol (10 mg/mL - clear, yellow solution), ethanol(50 mg/mL), acetone; very slightly soluble in chloroform; almost insoluble in water.

Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥98%
Colour of solution Passes
Identification Passes
Melting Temperature ≥258 °C
Loss on Drying ≤1.0%
Residue on Ignition ≤0.1%
Heavy Metals ≤0.0015%
Limit of Fluoran Passes
Chromatographic Purity Passes
Organic Volatile Impurities Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR