γ-(N-Morpholino)propanesulfonic acid (MOPS), white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 3-Morpholinopropane-1-sulfonic acid, 3-(N-Morpholino)propanesulfonic acid, 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

0210237010 0210237080 0210237083 0210237091 0210237025
IC10237010EA 42.83 USD
IC10237010 IC10237080 IC10237083 IC10237091 75875-590
γ-(N-Morpholino)propanesulfonic acid (MOPS), white powder
γ-(N-Morpholino)propanesulphonic acid (MOPS)

MOPS is a zwitterionic amino acid which acts as one of the "Good" buffers.


MOPS is used as a biological buffer.


Concentrations higher than 20 mM should not be used with mammalian cell cultures.


Soluble in water (to approximately 33% w/w - clear, colorless solution)


Caution: Research Use Only (RUO).

Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Melting Pt: 277 °C (lit.)
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
Merck Index: 13,06288
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=689465f1127e29c9dc1b03f99a64813530869406&vid=689465f1127e29c9dc1b03f99a64813530869406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
UV/Vis: (1M aq soln) OD250 nm ≤0.04
UV/Vis: (1M aq soln) OD280 nm ≤0.04
pH (10% aq soln) 3.2 - 4.2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR