D-(-)-Mannitol ≥96%, powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Mannitol
, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
02102248.5
0210224805
0210224801
IC10224801EA
90.44
USD
IC10224801
IC10224805
IC102248.5
D-(-)-Mannitol ≥96%, powder
D(-)-Mannitol
Mannitol is carbohydrate found in plants and plant exudates.
A sugar alcohol sweet tastant. Used in sweetness inhibition studies.
Soluble in water (1 g/5,5 ml), alcohol (1 g/83 ml), glycerol (1 g/18 ml); more soluble in hot water; soluble in pyridine, aniline, and aqueous solutions of alkalies; insoluble in ether.
Formula:
HOH₂C(CH(OH))₄CH₂OH MW: 182.17 g/mol Melting Pt: 163…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064287 CAS Number: 69-65-8 |
Specification Test Results
Synonyms | Manna sugar; Mannite |
Form | Powder |
Storage | Store at Room Temperature (15-30 °C). |
Loss on Drying | ≤1.0% |
Optical Rotation | +130 to +150°(c=1, ammonium molybdate, sulfuric acid) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...