Hydroxylammonium chloride ∼99%, powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Hydroxylamine hydrochloride
, Hydroxylamine HCl
0210199780
0210199790
IC10199790EA
203.02
USD
IC10199790
IC10199780
Hydroxylammonium chloride ∼99%, powder
Hydroxylammonium chloride
Soluble in water (100 mg/mL), alcohol, methanol, glycerol, propylene glycal or ethanol; insoluble in cold ether.
Formula:
NH₂OH·HCl MW: 69.49 g/mol |
MDL Number:
MFCD00051089 CAS Number: 5470-11-1 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...