Guanosine-5′-monophosphate 98, powder_white

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Guanosine 5'-Monophosphate hydrate, GMP, 5'-Guanylic acid, Guanosine 5'-monophosphoric acid

0210190880 0210190890 0210190891
IC10190880EA 58.01 USD
IC10190880 IC10190890 IC10190891
Guanosine-5′-monophosphate 98, powder_white
Guanosine-5′-monophosphate

Guanosine-5'-monophosphate is an ester of phosphoric acid with the nucleoside guanosine.


Guanosine-5′-monophosphate (GMP) has be used to study modulation of glutamatergic neurotransmission.


Soluble in water (5% clear solution)


Guanosine-5'-monophosphate is an ester of phosphoric acid with the nucleoside guanosine.


Caution: Research Use Only (RUO).

Formula: C₁₀H₁₄N₅O₈P
MW: 363.20 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00057066
CAS Number: 85-32-5

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Reference


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR