6-Aminocaproic acid, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
ε-Amino-n-caproic acid
, H-Eahx-OH
, Aminocaproic Acid
, 6-Aminocaproic Acid
, 6-Aminohexanoic acid
ε-Amino-n-Caproic Acid is a plasmin inhibitor. It inhibits carboxypeptidase B. It promotes rapid dissociation of plasmin by inhibiting the activation of plasminogen. As an inhibitor of plasmin it is has been utilized in the clotting buffer for fibrinogen assays.
EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxypeptidase, plasmin, and plasminogen activator. Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.
Soluble in water (50 mg/ml); sparingly soluble in alcohol and methanol; practically insoluble in ether; insoluble in ethanol.
Caution: Research Use Only (RUO).
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Melting Pt: 206,7 °C Density: 1,48 g/mL (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00008238 CAS Number: 60-32-2 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Melting Point | 205 - 215 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...