Dextran sulfate sodium salt, M.W. (6,000-8,000), white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Dextran sulfate sodium from Dextran, Dextran sulphate sodium from Dextran
Dextran sulfate is a polyanionic derivative of dextran produced by esterification of Dextran with chlorosulphonic acid. Dextran sulfate sodium (DSS) is the sodium salt which is a white to off-white powder freely soluble in water and salt solutions to form a stable, clear solution. The high purity and reproducible quality enables its application in pharmaceuticals and biotechnology industry.
Dextran Sulfate Sodium (DSS) which has proven to have versatile value in Biomedical/clinical research. DSS can be used as an anticoagulant, antiviral as well as antilipemic agent.
Antiviral and Antienzyme Activity – Baba et al., have shown in In Vitro studies that DSS inhibits the binding of various enveloped viruses including Human Immunodeficiency Virus (HIV), Herpes Virus (HSV) and Cytomegalovirus (CMV) to cells. Additionally, DSS has also shown to exhibit a potent inhibitory effect on many enzymes, for example hyaluronidase, adenylate cyclase, and amylase. Hofer at al., have shown that DSS induces bacterial translocation in HIV-infected humanized mice.
Solubility: Soluble in water (100 mg/ml - clear to slightly hazy yellow solution.
Caution: Research Use Only (RUO).
Formula:
(C₆H₇Na₃O₁₄S₃)n Storage Temperature: Ambient |
MDL Number:
MFCD00081551 CAS Number: 9011-18-1 |
Specification Test Results
Identity Test | Passes |
pH (1% aq soln) | 5.0 - 7.5 |
Specific Rotation | +75 ° to +105 ° |
Loss on Drying |
|
Residue on Ignition | 35 - 50% |
Molecular weight | Passes |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...