1-Hexadecanol, soluble

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Cetyl alcohol, Palmityl alcohol

0210130980 0210130990
IC10130980EA 50.12 USD
IC10130980 IC10130990
1-Hexadecanol, soluble
1-Hexadecanol

Easily soluble in diethyl ether; soluble in acetone,chloroform; slightly soluble in alcohol.


Cetyl alcohol is used in the cosmetic industry as an opacifier in shampoos, or as an emollient, emulsifier or thickening agent in the manufacture of skin creams and lotions. It is also employed as a lubricant for nuts and bolts.

Formula: H₃C(CH₂)₁₄CH₂OH
MW: 242.45 g/mol
MDL Number: MFCD00004760
CAS Number: 36653-82-4
Merck Index: 13,02037

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR