tert-Butyl-4-methoxyphenol (mixture of isomers) ∼98.5%, powder_white
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
tert-Butyl-4-methoxyphenol, BHA, tert-Butyl-4-hydroxyanisole, Butylhydroxyanisole, 2(3)-tert-Butyl-4-methoxyphenol, Butylated hydroxyanisole
Butylated Hydroxyanisole is an antioxidant. It is a mixture of 2- and 3-tertiary-butyl-4-hydroxyanisole.
Freely soluble in ethanol, methanol (50 mg/mL - clear and colorless) and in propylene glycol. Soluble in fats and oils. Insoluble in water
Butylated Hydroxyanisole is an antioxidant. It is a mixture of 2- and 3-tertiary-butyl-4-hydroxyanisole.
Caution: Research Use Only (RUO).
Formula:
C₁₁H₁₆O₂ MW: 180.25 g/mol Boiling Pt: 264 Flash Pt: 156 Storage Temperature: Ambient |
MDL Number:
MFCD01779059 CAS Number: 25013-16-5 Merck Index: 12,01582 |
Specification Test Results
Identity Test | Passes |
Purity | ~98.5% |
FTIR | Conforms to Reference |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...