5-Methylphenazinium methyl sulfate, dark yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Phenazine methosulfate, PMS, 5-Methylphenazin-5-ium methyl sulfate, 5-Methylphenazin-5-ium methyl sulphate, Phenazine methosulphate, Phenazine methylsulfate, Phenazine methylsulphate

0210095501 0210095505 0210095510
IC10095501EA 69.71 USD
IC10095501 IC10095505 IC10095510
5-Methylphenazinium methyl sulfate, dark yellow powder
5-Methylphenazinium methyl sulphate

Phenazine Methosulfate is an electron acceptor and carrier in enzyme systems. The oxidized form is yellow and the reduced form is colorless.


Phenazine methosulfate is used in tyrosine transaminase test and for enzymic determination of ethanol in blood by the colorimetric micromethod. Used with ascorbic acid to determine nitric oxide reductase activity. Since the reduced PMS is easily oxidized by oxygen, it is used in assays as an electron carrier between enzymes and oxygen, cytochrome c, indophenols, or tetrazolium salts. The reduced PMS is used as an electron donor to reduce cytochrome c or in photosynthetic experiments. Usage of PMS for detection of specific dehydrogenases has been reported.


PMS is reduced by flavoproteins such as succinic dehydrogenase. PMS is reduced non-enzymatically by NADH and NADPH. PMS is also reduced by dithionite, sodium borohydride, ascorbic acid, reduced ubiquinones, and reduced vitamin K. PMS reacts with sulfhydryl groups including those of enzymes.


Soluble in water(800 mg/4 mL), ethanol,methanol (5% w/v); insoluble in benzene, diethyl ether.


PMS is reduced by flavoproteins such as succinic dehydrogenase. PMS is reduced non-enzymatically by NADH and NADPH. PMS is also reduced by dithionite, sodium borohydride, ascorbic acid, reduced ubiquinones, and reduced vitamin K. PMS reacts with sulfhydryl groups including those of enzymes.

Formula: C₁₄H₁₄N₂O₄S
MW: 306.34 g/mol
Melting Pt: 198
MDL Number: MFCD00011923
CAS Number: 299-11-6
EINECS: 206-072-1
Merck Index: 12,06185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73a8e6522591723b5692a582242d68ad1938afe7&vid=73a8e6522591723b5692a582242d68ad1938afe7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR