L(+)-Asparagine, anhydrous ≥99%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: L-Asparagine, (S)-(+)-Asparagine, H-Asn-OH

0210079425 0210079480 0210079490
IC10079425EA 48.07 USD
IC10079425 IC10079480 IC10079490
L(+)-Asparagine, anhydrous ≥99%, white powder
L(+)-Asparagine

L-Asparagine is used in cell culture media and is a component of MEM non-essential amino acids solution. L-Asparagine has been shown to enhance ornithine decarboxylase activity in cultured human colon adenocarcinoma Caco-2 cells and in cultured IEC-6 intestinal epithelial cells. Spore germination in Bacillus subtilis has been increased in the presence of L-asparagine.


Soluble in aqueous solution (1% - clear, colorless solution).


L-Asparagine is used in cell culture media and is a component of MEM non-essential amino acids solution. L-Asparagine has been shown to enhance ornithine decarboxylase activity in cultured human colon adenocarcinoma Caco-2 cells and in cultured IEC-6 intestinal epithelial cells. Spore germination in Bacillus subtilis has been increased in the presence of L-asparagine.


Caution: Research Use Only (RUO).

Formula: H₂NCOCH₂CH(NH₂)CO₂H
MW: 132.12 g/mol
Boiling Pt: 343
Storage Temperature: Ambient
MDL Number: MFCD00064401
CAS Number: 70-47-3
Merck Index: 12,00872
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=3d09743d0834d64e4719e8295f634c39195c571f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Loss on Drying ≤1,0%
Optical Rotation (c=10, 1N HCl) +25° to +36°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR