L-Arginine hydrochloride, white crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
(S)-(+)-Arginine hydrochloride
L-arginine is an amino acid that is necessary for the production of protein, also helps rid the body of ammonia (a waste product) and stimulates the release of insulin. In addition, L-arginine is used to make nitric oxide (a compound that relaxes the blood vessels).
Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.
Soluble in water (50 mg/mL); slightly soluble in hot alcohol; insoluble in ether.
Caution: Research Use Only (RUO).
Formula:
C₆H₁₅ClN₄O₂ MW: 210.66 g/mol Melting Pt: 220…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064550 CAS Number: 1119-34-2 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Optical Rotation (c=8, 2 N HCl) | +21.0° to +23° |
Loss on Drying | ≤0.5% |
Melting Point | 220 - 230 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...