DL-α-Tocopherol acetate, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate

0210055525 0210055590 0210055580
IC10055525EA 45.2 USD
IC10055525 IC10055590 IC0210055580
DL-α-Tocopherol acetate, powder
DL-α-Tocopherol acetate

D-α-tocopherol is the predominant form of vitamin E in plasma and tissues. This product is a semi-synthetic acetate ester of naturally occuring α-tocopherol.


  • Presentation: Powder
  • Biochemical Category: Lipids and Vitamins


α-Tocopherol is a powerful inhibitor of the proliferation of estrogen receptor positive and estrogen receptor negative human breast cancer cell lines in a dose dependent manner in vitro.


Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens. Tocopherol acetate has properties similar but not identical to α-tocopherol.


Cut with corn starch and hydrolyzed protein.


D-α-tocopherol is the predominant form of vitamin E in plasma and tissues. This product is a semi-synthetic acetate ester of naturally occuring α-tocopherol.

Formula: C₃₁H₅₂O₃
Density: 0.96 g/mL (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00072042
CAS Number: 7695-91-2

Order Now

Specification Test Results

Identity Test Passes
Activity 250 u/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR