Space Gel, AIMS™
Supplier: AIMS
SPGL
95059-640EA 49.71 USD
95059-640
Space Gel, AIMS™
First Aid Supplies Surgical Drapes
The space gel is designed for use in the Thermo controlled surgery platform and the Thermo controlled recovery platform to warm lab animals during and after surgery.
The gels can be reused after reactivation.