Cardboard Freezer Boxes, Crystal Industries
Supplier: CRYSTAL TECHNOLOGY BE
MB2C-25 MB3C SB2C-100 SB2C-25 SB2C-49 SB2C-81 SB2CC-100 SB2CC-49 SB2CC-81 SB2CC-81-LP SB3C-100 SB3C-49 SB3C-81 SB4C SB4C-16 SB2C SB2CC-100W-LP
490000-772EA 15.28 USD
490000-772 490000-774 490000-804 490000-806 490000-808 490000-810 490000-812 490000-818 490000-820 490000-822 490000-826 490000-830 490000-832 490000-834 490000-836 490004-722 490004-721
Cardboard Freezer Boxes, Crystal Industries
Boxes Cryoboxes
A large variety of cardboard boxes are available with or without dividers.
  • All standard sizes available
  • With drain holes
  • Variety of cell dividers
Waterproof to survive repeated freezing and thawing.