Amikacin disulphate 50 mg/ml, sterile filtered

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulfate), (S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulphate)

J67496-AB J67496-AC
AAJ67496-ABEA 55.7 USD
AAJ67496-AB AAJ67496-AC
Amikacin disulphate 50 mg/ml, sterile filtered
Amikacin disulphate

Amikacin disulfate 50 mg/mL in distilled water and sterile-filtered.


  • Liquid


CAS No.: 39831-55-5 Storage Temperature: -30°C to -10°C Physical Form: Liquid MDL No.: MFCD00167475

Formula: C₂₂H₄₃N₅O₁₃·2H₂SO₄
MW: 781.77 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00167475
CAS Number: 39831-55-5
EINECS: 254-648-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR