Gentamicin sulfate 20 mg/mL in water, sterile filtered
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
NMD Inhibitor II
, Garamycin
, Gentiomycin C
, Gentamicin sulfate salt
, Gentamicin sulphate salt
J67488-AD
AAJ67488-ADEA
316.48
USD
AAJ67488-AD
Gentamicin sulfate 20 mg/mL in water, sterile filtered
Gentamicin sulphate
Storage Temperature:
Refrigerator |
MDL Number:
MFCD00270181 CAS Number: 1405-41-0 EINECS: 215-778-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...