Spectinomycin dihydrochloride pentahydrate 50 mg/mL in water

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate

J67387-8EQ J67387-XF
AAJ67387-8EQEA 353 USD
AAJ67387-8EQ AAJ67387-XF
Spectinomycin dihydrochloride pentahydrate 50 mg/mL in water
Spectinomycin dihydrochloride pentahydrate

Spectinomycin have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr.


This antibiotic is used to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons. It is recommended for use in cell culture applications at 7.5 to 20 mg/L.


Fully miscible in water. Soluble in methanol, DMSO (~11 mg/ml), ethanol (~0.3 mg/ml), and DMF (~2 mg/ml).


Caution: Protect from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Formula: C₁₄H₂₄N₂O₇·2HCl·5H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00150886
CAS Number: 22189-32-8
EINECS: 244-554-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR