Penicillin G potassium salt 100 mg/mL in distilled water, sterile filtered
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt
J67333-AD
J67333-AE
AAJ67333-ADEA
76.7
USD
AAJ67333-AD
AAJ67333-AE
Penicillin G potassium salt 100 mg/mL in distilled water, sterile filtered
Penicillin G potassium salt
Penicillin G potassium salt, 100 mg/ml in distilled water, sterile-filtered
Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₁₆H₁₇KN₂O₄S MW: 372.49 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00036193 CAS Number: 113-98-4 EINECS: 204-038-0 Merck Index: 12,07225 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...