Streptomycin sulfate 100 mg/ml in distilled water, sterile filtered
Supplier: Thermo Scientific Chemicals
|
Warning
|
J67224-AD
J67224-AE
AAJ67224-ADEA
154.55
USD
AAJ67224-AD
AAJ67224-AE
Streptomycin sulfate 100 mg/ml in distilled water, sterile filtered
Streptomycin sulphate
Streptomycin sulfate, 100 mg/mL in distilled water, sterile-filtered.
Liquid
Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₂₁H₃₉N₇O₁₂·1.5H₂SO₄ MW: 1457.4 g/mol Melting Pt: 250 Storage Temperature: Refrigerator |
MDL Number:
MFCD00037023 CAS Number: 3810-74-0 EINECS: 223-286-0 Merck Index: 12,08983 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...