Pyrazinamide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Pyrazinecarboxamide, Pyrazine-2-carboxamide, 2-Pyrazinecarboxamide

J66822-09 J66822-14 J66822-22
10067-286EA 44.7 USD
10067-286 10067-288 10067-410
Pyrazinamide 98%
Pyrazinamide

Pyrazinamide, 98%


Purity: 98% CAS No.: 98-96-4 Molecular Formula: C5H5N3O Formula Weight: 123.11 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White Odor: Odorless Melting Point: 189-191° Solubility: Soluble in water Beilstein Reference: 112306 MDL No.: MFCD00006132

Formula: C₅H₅N₃O
MW: 123.11 g/mol
Melting Pt: 189…191 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006132
CAS Number: 98-96-4
EINECS: 202-717-6
Merck Index: 14,7956

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR