oxolinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

J66637-06 J66637-14
10067-230EA 162 USD
10067-230 10067-232
oxolinic acid 98%
oxolinic acid

Targets Gram-negative bacteria


Purity: 98% CAS No.: 14698-29-4 Molecular Formula: C13H11NO5 Formula Weight: 261.23 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to off-white Melting Point: 314-316° dec. Boiling Point: 473° Solubility: Soluble in 0.5N NaOH with warming Beilstein Reference: 620635 MDL No.: MFCD00056775


Applications: Targets gram negative bacteria

Formula: C₁₃H₁₁NO₅
MW: 261.233 g/mol
Boiling Pt: 473 °C
Melting Pt: 314…316 °C (dec.)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00056775
CAS Number: 14698-29-4
EINECS: 238-750-8
Merck Index: 14,6965

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR