Propidium iodide 1mg/ml in aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide

J66584-AB
AAJ66584-ABEA 180.23 USD
AAJ66584-AB
Propidium iodide 1mg/ml in aqueous solution
Propidium iodide

Applications: Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Selectively stains dead cells and a counter stain for multicolor fluorescent images.

Formula: C₂₇H₃₄I₂N₄
MW: 668.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011921
CAS Number: 25535-16-4
EINECS: 247-081-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR