Ampicillin trihydrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate

J66514-14 J66514-22 J66514-36
10067-182EA 85.2 USD
10067-182 10067-184 10067-186
Ampicillin trihydrate
Ampicillin trihydrate

Ampicillin is lactam antibiotic with an amino group side chain attached to the penicillin structure


White powder


It is commonly used to select for ampicillin resistance in mutated and transformed cells.


It inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Effective on Gram-negative and Gram-positive bacteria.


Soluble in water. Practically insoluble in ethanol


Caution: Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: C₁₆H₁₉N₃O₄S·3H₂O
MW: 403.45 g/mol
Boiling Pt: 684 °C
Melting Pt: 208 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00072036
CAS Number: 7177-48-2
EINECS: 200-709-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR