Sparfloxacin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Amino-1-cyclopropyl-7-((3S,5R)-3,5-dimethylpiperazin-1-yl)-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

J66358-03 J66358-09
10067-136EA 36 USD
10067-136 10067-402
Sparfloxacin 98%
Sparfloxacin

Targets a wide range of Gram-positive, Gram-negative, and mycoplasma species


Applications: Targets a wide range of gram positive, gram negative, and mycoplasma species


Applications: Targets a wide range of gram positive, gram negative, and mycoplasma species

Formula: C₁₉H₂₂F₂N₄O₃
MW: 392.407 g/mol
Boiling Pt: 640 °C
Melting Pt: 265 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00869619
CAS Number: 110871-86-8
Merck Index: 14,8735

Order Now

Specification Test Results

Assay 98.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR