Tween® 20 (Polysorbate), Liquid Electrophoresis

Supplier: Thermo Scientific Chemicals

Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated

J66278-AE J66278-AP
AAJ66278-AEEA 28.99 USD
AAJ66278-AE AAJ66278-AP
Tween® 20 (Polysorbate), Liquid Electrophoresis
Polysorbate 20

Polysorbate 20, Electrophoresis Reagent


CAS No.: 9005-64-5 Molecular Formula: C58H11O26 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.105 Refractive Index: 1.4700 Appearance: Colorless to pale yellow Boiling Point: 100° MDL No.: MFCD00165986

Formula: C₅₈H₁₁₄O₂₆
Boiling Pt: >100 °C
Density: 1.105
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR