3B Scientific® Head And Neck Musculature Models
Supplier: AMERICAN 3B SCIENTIFIC SE
1001239
470119-046EA 416.85 USD
470119-046
3B Scientific® Head And Neck Musculature Models
Models Human Biology Models
These head and neck models all begin with a representation of the superficial musculature of head and neck showing: Parotid gland, Submandibular gland (right half), Deep musculature (left half), Lower jaw partially exposed.
  • Includes 3B Smart Anatomy
Additionally you can choose to show either the blood vessels, or the nerves. Get all three for comparison, or just the one you need.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.