Life/form® Ostomy Care Simulator
Supplier: NASCO HEALTHCARE INC. SE
LF00906
470119-380EA 1303.25 USD
470119-380
Life/form® Ostomy Care Simulator
Models Human Biology Models
Introduce Ostomy Care To Students And Patients
  • Demonstrate and practice colostomy and ileostomy
  • Practice dilation and cleaning
  • Includes carrying case
Reproduction of an abdomen showing the anatomy of both a colostomy and ileostomy to provide lifelike functions and appearance. Dilation of the stomas can be demonstrated and practiced, along with application of postoperative and permanent ostomy bags. The colostomy can be irrigated. Drainage and excretion at the ileostomy (water) and colostomy (simulated stool) can be pumped by syringes.
Delivered with simulated stool and carrying case.