Laerdal® Blood Pressure Training Arm
Supplier: AMERICAN 3B SCIENTIFIC SE
1017977
470119-354EA 2541.59 USD
470119-354
Laerdal® Blood Pressure Training Arm
Models Human Biology Models
Lifelike, Adult Blood Pressure Training Arm
  • Palpable antecubital pulse
  • Blood pressure trainer with LCD guided operation
  • Systolic, diastolic, heart rate and auscultatory gap are programmable
  • Representation of both systolic and diastolic pressures
  • Indication of gauge reading as pressure is increased or decreased
  • Adjustable volume
This blood pressure training arm is complete with a palpable antecubital pulse. Features of this blood pressure arm include:. Blood Pressure Trainer with LCD guided operation. Systolic, diastolic, heart rate and auscultatory gap are programmable. Representation of both systolic and diastolic pressures. Indication of gauge reading as pressure is increased or decreased. Volume adjustable. Includes blood pressure trainer, cuff, 9-volt battery and carry case. This simulator is a great way to teach and learn the technique of taking a patients blood pressure.