SP Bel-Art Ultra-Low 5-Box Freezer Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
H18992-0012 H18992-0000 H18992-0022 H18992-0002
10154-404CS 1048.8 USD
10154-404 10154-406 10154-400 10154-402
SP Bel-Art Ultra-Low 5-Box Freezer Racks, Bel-Art Products, a part of SP
Racks Cryogenic Box Racks
Easily configured to customize freezer space, these modular ultra low freezer racks link together side-by-side or can be securely stacked for horizontal and/or vertical storage.
  • Modular design is easy to customize
  • Stackable
  • Optional top cover
  • Lightweight
Purchase as many or as few as is needed for your specific freezer space or storage needs. Includes pull out drawer for easy sample retrieval even from the back of the freezer. Each lightweight, wire formed rack holds 5 freezer boxes up to 5¼ x 5¼ x 2”H. Available in 4 colors for easy sample identification. An optional top cover provides flat shelf space for additional storage.