Micro Tubes with Thread, Type A, Sarstedt
Supplier: Sarstedt
72.730.003 72.730.307 72.703.406 72.730.308 72.730.305 72.730.306 72.730.309 72.730
101093-797PK 51.43 USD
101093-797 101093-589 101093-779 101093-616 102971-720 103575-128 101093-597 101093-740
Micro Tubes with Thread, Type A, Sarstedt
Tubes Microtubes
Micro Tubes are for transport, storage and sample processing in the laboratory with safe screw cap with O-ring.
  • Autoclavable at 121 °C
  • Tubes are skirted for self-standing and neutral cap
Knurls enable single-handed operation in special Sarsetedt racks for micro tubes.