Anti-CISD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16006-1-AP-150UL
10084-600EA 462.47 USD
10084-600
Anti-CISD1 Rabbit Polyclonal Antibody
Antibodies
MitoNEET, also named CISD1, belongs to a previously uncharacterized ancient family of proteins for which the hallmark is the presence of a unique 39 amino acid CDGSH domain. It is a single-pass type III membrane protein, located in mitochondrion outer membrane and may play a role in regulating maximal capacity for electron transport and oxidative phosphorylation. MitoNEET is a recently identified drug target for a commonly prescribed diabetes drug, Pioglitazone. This antibody recognizing MitoNEET (calculated 12 kDa) as a 17 kDa protein may be due to its posttranslational modification or metal binding activity.

Western Blot: Mouse Skeletal Muscle Tissue, 1:1000-1:10000; IHC: Human Heart Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: HepG2 Cells, 1:1000-1:10000; FC: HeLa Cells, N/A

Type: Primary
Antigen: CISD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR