Magnetic Lines Viewer
Supplier: AMERICAN EDUCATIONAL PROD
7-1257
470117-484EA 23.75 USD
470117-484
Magnetic Lines Viewer
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
For classroom demonstrations
  • Great for overhead projectors
  • 7/16 x 4 3/4 x 8 3/4"; .64Ib
Great visibility makes understanding the lines of magnetic force much easier for your students! Iron filings float in a layer of oil in an acrylic plate and orient themselves according to the magnetic force of any magnet placed under the apparatus.