Caps for NMR Sample Tubes, Chemglass
Supplier: Chemglass
CG-1842-10 CG-1842-01 CG-1842-02 CG-1842-03 CG-1842-04 CG-1842-05 CG-1842-06
80061-568EA 104.15 USD
80061-568 80061-556 80061-558 80061-560 80061-562 80061-564 80061-566
Caps for NMR Sample Tubes, Chemglass
Closures Test/Sample Tube Closures
Made from polyethylene, these caps are sized to fit 5mm O.D
NMR sample tubes. Cap fits tightly to stay in place during increased internal vaporization pressure. Available in a pack of 100, or in a convenient divided polypropylene box having 100 each of green, red, blue, yellow, white, and black caps.