May Grunwald's eosin-methylene blue, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ400

1.01352.0025 1.01352.0100 1.01352.0025 1.01352.0100
EM1.01352.0025CS 2667.88 USD
CA1.01352.0025 CA1.01352.0100 EM1.01352.0025 EM1.01352.0100
May Grunwald's eosin-methylene blue, Sigma-Aldrich®
May Grunwald's eosin-methylene blue
Storage Temperature: Ambient
MDL Number: MFCD00131580
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR