Reli® Quincke Needles, Sterile, MYCO Medical
Supplier: MYCO Medical
SN22G151-RW SN25G151 SN23G351 SN22G801-SB SN23G601 SN20G501 SN22G351-SB SN25G251 SN22G701-RW SN20G601 SN22G501-RW SN25G801 SN19G351 SN23G501 SN27G351 SN22G801-RW SN25G501 SN25G701 SN18G301 SN26G351 SN25G601 SN22G251 SN22G501-SB SN25G351 SN18G501 SN18G351 SN25G301 SN20G351 SN22G601-RW SN22G351
10148-734BX 133.58 USD
10148-734 10148-712 10148-716 10148-738 10148-616 10148-718 10148-618 10148-620 10148-622 10148-688 10148-720 10148-742 10148-708 10148-624 10148-744 10148-722 10148-700 10148-626 10148-702 10148-726 10148-704 10148-706 10148-692 10148-694 10148-730 10148-696 10148-710 10148-732 10148-698 10148-690
Reli® Quincke Needles, Sterile, MYCO Medical
Syringe Needles
Reli Quincke Needles are high quality needles with large, clear, color-coded hubs for enhanced tactile feel and easy gauge identification
Matched-Point grinding allows for tight tolerance between the stylet and cannula to provide for smooth, low-force insertion. Reli Quincke Needles are available in a broad range of sizes for maximum flexibility.