Triethanolamine (trolamine), EMPLURA®, Supelco®

Supplier: MilliporeSigma

Synonyms: 2,2',2''-NitrilotriethanolTris-(2-hydroxyethyl)amine

M-Clarity™ quality level = MQ200

8.22341.9026 8.22341.9026
EM8.22341.9026EA 737.48 USD
CA8.22341.9026 EM8.22341.9026
Triethanolamine (trolamine), EMPLURA®, Supelco®
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 335.4 °C (1013 hPa)
Melting Pt: 21.6 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 179 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 12,09798
https://us.vwr.com/stibo/web/std.lang.all/75/12/2297512.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR