N-Methyl-2-pyrrolidone (NMP), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: N-MethylpyrrolidinoneN-methylpyrrolidoneN-Methyl-gamma-butyrolactamN-Methyl-2-pyrrolidinoneM-PyrolN-Methyl-alpha-pyrrolidone1-Methyl-2-pyrrolidoneNMPMethyl PyrrolidoneN-Methyl-2-pyrrolidoneN-Methyl-2-pyrrolidinone,NMPN-Methylpyrrolidone1-Methyl-2-pyrrolidinone

M-Clarity™ quality level = MQ200

8.06072.1000 8.06072.9011 8.06072.1000 8.06072.2500 8.06072.9025 8.06072.9011 8.06072.2500 8.06072.9025
EM8.06072.2500CS 1951 USD
EM8.06072.2500 EM8.06072.9011 CA8.06072.1000 CA8.06072.9011 CA8.06072.2500 CA8.06072.9025 EM8.06072.9025 EM8.06072.1000
N-Methyl-2-pyrrolidone (NMP), EMPLURA®, Supelco®
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 12,06197
https://us.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR