Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide
M-Clarity™ quality level = MQ200
1.04971.1000
1.04971.9050
EM1.04971.1000CS
5021
USD
EM1.04971.1000
EM1.04971.9050
Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), Supelco®
Potassium hexacyanoferrate (III)
Formula:
K₃Fe(CN)₆ MW: 329.25 g/mol Density: 1.85 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011392 CAS Number: 13746-66-2 EINECS: 237-323-3 UN: 3077 ADR: 9 III Merck Index: 13,07712 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...