Potassium thiocyanate, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Potassium rhodanide

M-Clarity™ quality level = MQ200

1.05124.1000 1.05124.9050
EM1.05124.1000CS 1035 USD
EM1.05124.1000 EM1.05124.9050
Potassium thiocyanate, Supelco®
Potassium thiocyanate
Formula: KSCN
MW: 97.18 g/mol
Boiling Pt: 500 (dec.)
Melting Pt: 175
Density: 1.91 g/cm3 at 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR