Antimony(III) potassium oxitartrate trihydrate ≥99.0%, EMPLURA®, Extra Pure, Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.08092.0250 1.08092.1000
EM1.08092.0250CS 1873 USD
EM1.08092.0250 EM1.08092.1000
Antimony(III) potassium oxitartrate trihydrate ≥99.0%, EMPLURA®, Extra Pure, Supelco®
Antimony (III) potassium oxitartrate trihydrate
Formula: C₈H₄K₂O₁₂Sb₂·3H₂O
Melting Pt: 100 °C
Density: 2.6 g/cm³ (20 °C)
MDL Number: MFCD00167056
CAS Number: 28300-74-5
EINECS: 229-436-1
UN: 1551
ADR: 6.1,III
Merck Index: 13,00706

Order Now

Specification Test Results

Assay (iodometric) 99.0 - 103.0 %
Identity passes test
As (Arsenic) ≤ 0.015 %
Pb (Lead) ≤ 0.002 %
Loss on drying ≤ 2.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR