Isobutanol ≥98.5% (by GC), Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
2-Methyl-1-propanol
, Isobutylalcohol
, iso-Butanol
, iso-Butyl alcohol
M-Clarity™ quality level = MQ200
1.00985.6025
1.00985.2500
1.00985.9190
1.00985.2500
1.00985.6025
1.00985.9190
EM1.00985.2500CS
738.51
USD
EM1.00985.2500
CA1.00985.9190
CA1.00985.6025
CA1.00985.2500
EM1.00985.9190
EM1.00985.6025
Isobutanol ≥98.5% (by GC), Supelco®
Isobutanol
Formula:
(CH₃)₂CHCH₂OH MW: 74.12 g/mol Boiling Pt: 106…108 °C (1013 hPa) Melting Pt: –108 °C Density: 0.802 g/cm3 (20 °C) Flash Pt: 28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004740 CAS Number: 78-83-1 EINECS: 201-148-0 UN: 1212 ADR: 3,III Merck Index: 13,05148 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...