Formamide, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Carbamaldehyde

M-Clarity™ quality level = MQ200

1.04008.1000 1.04008.2500 1.04008.9025
EM1.04008.1000CS 2173 USD
EM1.04008.1000 EM1.04008.2500 EM1.04008.9025
Formamide, EMPLURA®, Supelco®
Formamide
Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR