Petroleum spirit, 40…80 °C for denaturation, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Petroleum ether

M-Clarity™ quality level = MQ200

1.01769.6025 1.01769.9190 1.01769.6010 1.01769.5000 1.01769.9190 1.01769.6025 1.01769.6010 1.01769.5000 1.01769.1000 1.01769.1000
EM1.01769.1000CS 520 USD
CA1.01769.9190 CA1.01769.6025 CA1.01769.1000 CA1.01769.5000 EM1.01769.1000 EM1.01769.9190 CA1.01769.6010 EM1.01769.5000 EM1.01769.6025 EM1.01769.6010
Petroleum spirit, 40…80 °C for denaturation, Supelco®
Petroleum spirit
Boiling Pt: >50 °C (1013 hPa)
Density: 0.666 g/cm3 (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR