Petroleum spirit, 50…70 °C, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Petroleum ether
M-Clarity™ quality level = MQ200
1.00910.6025
1.00910.1000
1.00910.6025
1.00910.1000
1.00910.5000
1.00910.5000
EM1.00910.1000CS
653
USD
EM1.00910.1000
EM1.00910.5000
EM1.00910.6025
CA1.00910.5000
CA1.00910.6025
CA1.00910.1000
Petroleum spirit, 50…70 °C, Supelco®
Petroleum spirit
Boiling Pt:
50…70 °C (1013 hPa) Melting Pt: < –20 °C Density: 0.654…0.67 g/cm³ (20 °C) Flash Pt: <–21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 265-151-9 UN: 1268 ADR: 3,II |